Skip to Content
MilliporeSigma
All Photos(1)

Documents

Z509302

Sigma-Aldrich

Ultrasonic processors 500W and 750W

750W, AC/DC input 110 V AC

Synonym(s):

sonicator, ultra sonicator, ultrasonic homogenizer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41101519

manufacturer/tradename

Sonics & Materials VCX750-110V

availability

not available in EU

Looking for similar products? Visit Product Comparison Guide

General description

The VCX 750 is a powerful and versatile ultrasonic liquid processor. Using a variety of accessories, the system can process a wide range of sample types and volumes for many different applications.The VCX 750 is microprocessor based and programmable. Intuitive screens are user friendly and easy to navigate. The 750 watt power supply has the capability of operating our largest assortment of horns and accessories. A standard 1/2" diameter probe (50 – 250mL volumes) with replaceable tip is included with the system.



  • Automatic tuning and frequency control
  • Integrated temperature controller from 1°C to 100°C
  • Stores up to 10 procedures
  • Real time display
  • Control process time from 1 second to 10 hours
  • Shipped complete with a 1/2 in. (13 mm) probe (630-0220) with replaceable tip, tool kit, and instruction manual

Applications:
Cell disruption in bacteria, spores, and tissues; accelerating catalytic reactions; extractions of serums, toxins, enzymes, and viruses from organic sources; solubilizing difficult compounds; emulsification to 0.01 mm; homogenizing liquids; sample preparation from microscopy and particle size analysis.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Surfactant-assisted dispersion of single-walled carbon nanotubes for debundling or exfoliation in dispersion procedures.

Surfactant-assisted dispersion of single-walled carbon nanotubes for debundling or exfoliation in dispersion procedures.

Surfactant-assisted dispersion of single-walled carbon nanotubes for debundling or exfoliation in dispersion procedures.

Surfactant-assisted dispersion of single-walled carbon nanotubes for debundling or exfoliation in dispersion procedures.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service