Skip to Content
MilliporeSigma
All Photos(1)

Documents

Z503967

Laboratory brushes with PP handles

Beaker, brush diam. × L 2 1/2 in. × 6 in.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47131605
NACRES:
NB.35

Overall L

13 in.

brush diam. × L

2 1/2 in. × 6 in.

Looking for similar products? Visit Product Comparison Guide

General description

Durable black nylon filament brushes with tufted ends have white, textured-grip, molded PP handles. Brushes will not scratch or mar equipment, absorb moisture, chemicals, or oils.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service