Skip to Content
MilliporeSigma
All Photos(1)

Documents

Z281336

Laboratory brushes

Funnel, L 13 in., brush diam. × L 2 1/4 in. × 6 in.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47131605
NACRES:
NB.35

packaging

pkg of 4 ea

L

13 in.

brush diam. × L

2 1/4 in. × 6 in.

Looking for similar products? Visit Product Comparison Guide

General description

Brush materials resistant to acids, aromatic and aliphatic hydrocarbons, ketones, ethyl acetate esters, trichloroethylene, and hot water. FDA and USDA approved materials.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service