Documents
Z281379
Laboratory brushes
Burette, L 36 in., brush diam. × L 3/4 in. × 3 1/4 in.
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
packaging
pkg of 4 ea
L
36 in.
brush diam. × L
3/4 in. × 3 1/4 in.
General description
Brush materials resistant to acids, aromatic and aliphatic hydrocarbons, ketones, ethyl acetate esters, trichloroethylene, and hot water. FDA and USDA approved materials.