Key Documents
BAF170680000
Cleanware swivel head bottle/flask brush
pack of 1 ea
Synonym(s):
bottle brush
About This Item
UNSPSC Code:
41103200
NACRES:
NB.35
Pricing and availability is not currently available.
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F17068-0000
General description
Use this brush to clean any type of deep vessel. Good for restaurant kitchens, laboratories with round-bottom flasks, repair shops, any facility needing dexterity and small size in the cleaning head.
Diameter of wooden head: 1.7025 in. x .8915 in. and the bristles are 1.578 in. x 0.74 in.
  • Head of brush swivels, reaching intricate curves and corners
  • Natural bristles set in wood will not scrape cleaned surface
  • Head fits into any container with a neck diameter of 24 mm (1 in.) or more.
  • Reach into deep vessels with 33 cm (13 in.) hardwood handle
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)