Documents
Z190535
Supelco
Liquid scintillation vials (with screw cap attached)
white polypropylene cap, metal foil liner, pkg of 500 ea
Synonym(s):
Scintillation Counting Vials
About This Item
UNSPSC Code:
41121806
NACRES:
NB.11
material
clear glass vial
metal foil liner
white polypropylene cap
feature
closure type screw top vial
closure type white polypropylene cap
packaging
pkg of 500 ea
size
28 mm × 61 mm
volume
20 mL
fitting
thread for 22-400
General description
Autoclavable, easy to maintain. A vial is used to store or as a closure type,in order to prevent contamination.
Reliable Wheaton "180" brand low-potassium glass. White 22 mm caps have tops suitable for writing. Vials with caps attached are packed in individual cells in trays of 100.
Visit the Document Library
Laura G Wilkinson et al.
Plant methods, 13, 67-67 (2017-08-22)
Seed development in the angiosperms requires the production of a female gametophyte (embryo sac) within the ovule. Many aspects of female reproductive development in cereal crops are yet to be described, largely due to the technical difficulty in obtaining phenotypic