Skip to Content
MilliporeSigma
All Photos(1)

Documents

W326909

Sigma-Aldrich

3,5-Dimethyl-1,2-cyclopentadione

≥97%

Synonym(s):

3,5-dimethylcyclopentane-1,2-dione, Caramel dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10O2
CAS Number:
Molecular Weight:
126.15
FEMA Number:
3269
EC Number:
Council of Europe no.:
2234
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.076
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

Halal
Kosher

Assay

≥97%

mp

87-95 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

caramel; maple; sweet

storage temp.

2-8°C

SMILES string

CC1CC(C)C(=O)C1=O

InChI

1S/C7H10O2/c1-4-3-5(2)7(9)6(4)8/h4-5H,3H2,1-2H3

InChI key

MIDXCONKKJTLDX-UHFFFAOYSA-N

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service