Skip to Content
MilliporeSigma
All Photos(1)

Documents

W330507

Sigma-Aldrich

3-Methyl-1,2-cyclohexanedione

≥98%, FG

Synonym(s):

coffee dione

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H7(=O)2
CAS Number:
Molecular Weight:
126.15
FEMA Number:
3305
EC Number:
Council of Europe no.:
2311
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.080
NACRES:
NA.21

biological source

synthetic

grade

FG
Halal

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002

Assay

≥98%

bp

69-72 °C/1 mmHg (lit.)

mp

61-63 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

bread; burnt; caramel; coffee; maple

SMILES string

CC1CCCC(=O)C1=O

InChI

1S/C7H10O2/c1-5-3-2-4-6(8)7(5)9/h5H,2-4H2,1H3

InChI key

JDXJKLGWPNXSHL-UHFFFAOYSA-N

Gene Information

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service