Skip to Content
MilliporeSigma
All Photos(1)

Documents

W334006

Sigma-Aldrich

2-Ethoxythiazole

≥99%, FG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H7NOS
CAS Number:
Molecular Weight:
129.18
FEMA Number:
3340
EC Number:
Council of Europe no.:
11611
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
15.021
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 872/2012

Assay

≥99%

refractive index

n20/D 1.504 (lit.)

bp

157-160 °C (lit.)

density

1.133 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

musty; vegetable; nutty

storage temp.

2-8°C

SMILES string

CCOc1nccs1

InChI

1S/C5H7NOS/c1-2-7-5-6-3-4-8-5/h3-4H,2H2,1H3

InChI key

NDUWJHRKDYXRAD-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

129.2 °F - closed cup

Flash Point(C)

54 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service