Skip to Content
MilliporeSigma

A8176

N-Acetyl-D-mannosamine

≥98% (TLC)

Synonym(s):

N-acetylmannosamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C8H15NO6
CAS Number:
Molecular Weight:
221.21
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

animal (crustaceans)

Quality Segment

assay

≥98% (TLC)

form

powder

technique(s)

thin layer chromatography (TLC): suitable

color

white to off-white

solubility

water: 50 mg/mL, clear, colorless to faintly yellow

storage temp.

−20°C

SMILES string

CC(=O)N[C@@H]1[C@@H](O)O[C@H](CO)[C@@H](O)[C@@H]1O

InChI

1S/C8H15NO6/c1-3(11)9-5-7(13)6(12)4(2-10)15-8(5)14/h4-8,10,12-14H,2H2,1H3,(H,9,11)/t4-,5+,6-,7-,8+/m1/s1

InChI key

OVRNDRQMDRJTHS-UOLFYFMNSA-N

Application

N-Acetyl-D-mannosamine (ManNAc) is used as a substrate to identify, differentiate and characterize enzymes such as N-acetyl-glucosamine epimerase(s) and aldolase(s). N-Acetyl-D-mannosamine is used for the synthesis of of N-acetylneuraminic acid.

Biochem/physiol Actions

N-Acetyl-D-mannosamine (ManNAc) is an essential precursor of N-acetylneuraminic acid (NeuAc), the specific monomer of polysialic acid. Transport studies in the E. coli K1 strain have been performed to probe the effect of ManNAc on capsular polysialic acid production.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.


Still not finding the right product?

Explore all of our products under N-Acetyl-D-mannosamine


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library