Skip to Content
MilliporeSigma
All Photos(1)

Documents

W309702

Sigma-Aldrich

Undecyl alcohol

97%, FG

Synonym(s):

1-Undecanol, Alcohol C11, Undecyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)10OH
CAS Number:
Molecular Weight:
172.31
FEMA Number:
3097
Beilstein:
1698334
EC Number:
Council of Europe no.:
751
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.057
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.515

Assay

97%

refractive index

n20/D 1.44 (lit.)

bp

146 °C/30 mmHg (lit.)

mp

11 °C (lit.)

density

0.83 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fresh; waxy; soapy

SMILES string

CCCCCCCCCCCO

InChI

1S/C11H24O/c1-2-3-4-5-6-7-8-9-10-11-12/h12H,2-11H2,1H3

InChI key

KJIOQYGWTQBHNH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Undecyl alcohol has been reported as a key volatile constituent of Senecio tenuifolius and Flacourtia montana.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

226.4 °F - closed cup

Flash Point(C)

108 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Volatile Constituents of Senecio tenuifolius From India.
Joshi, R. K
Chemistry of Natural Compounds, 52(5), 926-927 (2016)
Chemical analysis of volatile constituents of Flacourtia montana j. Graham (salicaceae): an edible and wild fruit of western ghats of india.
Mundaragi AC, et al.
Plant Archives, 17(1), 506-508 (2017)
Fateme Mirrahimi et al.
Journal of AOAC International, 97(3), 933-937 (2014-07-24)
A sensitive and selective method for the determination of low levels of rhodium (Rh) in environmental samples is needed. In the proposed method, an extracting solvent with a lower toxicity and density than the other solvents typically used in dispersive
Xiaohong Hou et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 969, 123-127 (2014-08-30)
A novel ultrasound-assisted dispersive liquid-liquid microextraction based on solidification of floating organic droplet method (UA-DLLME-SFO) combined with gas chromatography (GC) was developed for the determination of eight pyrethroid pesticides in tea for the first time. After ultrasound and centrifugation, 1-dodecanol
Yuzhen Guo et al.
Lab on a chip, 15(7), 1759-1764 (2015-02-17)
A simple method to fabricate microchannels is demonstrated based on an inkjet printing liquid template. The morphology of the liquid template can be well controlled by using ink with viscosity sensitive to temperature. The as-prepared Y-shape microchannel is used as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service