Skip to Content
MilliporeSigma
All Photos(1)

Documents

W371009

Sigma-Aldrich

Methyl 3-nonenoate

≥96%, FG

Synonym(s):

Methyl non-3-enoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CH=CHCH2CO2CH3
CAS Number:
Molecular Weight:
170.25
FEMA Number:
3710
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.298
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002

Assay

≥96%

refractive index

n20/D 1.436 (lit.)

density

0.885 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; fruity; pear

SMILES string

CCCCC\C=C\CC(=O)OC

InChI

1S/C10H18O2/c1-3-4-5-6-7-8-9-10(11)12-2/h7-8H,3-6,9H2,1-2H3/b8-7+

InChI key

MTDCXFZGUVZRSQ-BQYQJAHWSA-N

General description

Natural occurence: Guava

Application

This interesting material has notes that make it an obvious choice for cucumber and melon flavors. Beyond that, however, it will also work in other fruits, like pear, guava, peach, and some mint flavors, especially spearmint. It might even be considered at very low levels for use in cheese flavors, especially those from goat and sheep′s milk.

Biochem/physiol Actions

Taste at 3-6ppm

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

190.4 °F - closed cup

Flash Point(C)

88 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organoleptic Characteristics of Flavor Materials
Michalski, J.
Perfumer & Flavorist, 38(1), 47-47 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service