Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

289485

Sigma-Aldrich

3,5,5-Trimethyl-1-hexanol

technical grade, 85%

Synonym(s):

3,5,5-Trimethyl-1-hexanol, 3,5,5-Trimethylhexanol, 3,5,5-Trimethylhexyl alcohol, Nonylol, i-Nonyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CCH2CH(CH3)CH2CH2OH
CAS Number:
Molecular Weight:
144.25
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

85%

form

liquid

refractive index

n20/D 1.432 (lit.)

bp

193-194 °C (lit.)

density

0.824 g/mL at 25 °C (lit.)

SMILES string

CC(CCO)CC(C)(C)C

InChI

1S/C9H20O/c1-8(5-6-10)7-9(2,3)4/h8,10H,5-7H2,1-4H3

InChI key

BODRLKRKPXBDBN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A dermatologic and toxicologic analysis of 3,5,5-trimethyl-1-hexanol (when used as a fragrance ingredient) was reported.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT RE 2 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

176.0 °F - closed cup

Flash Point(C)

80 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D McGinty et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 48 Suppl 4, S47-S50 (2010-08-13)
A toxicologic and dermatologic review of 3,5,5-trimethyl-1-hexanol when used as a fragrance ingredient is presented. 3,5,5-Trimethyl-1-hexanol is a member of the fragrance structural group branched chain saturated alcohols. The common characteristic structural elements of the alcohols with saturated branched chain

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service