Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

W242217

Sigma-Aldrich

Ethyl benzoate

greener alternative

natural, ≥99%, FCC, FG

Synonym(s):

Benzoic acid ethyl ester, Benzoic ether, Ethyl benzene carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COOC2H5
CAS Number:
Molecular Weight:
150.17
FEMA Number:
2422
Beilstein:
1908172
EC Number:
Council of Europe no.:
261
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.726
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
natural
food allergen:
no known allergens
Pricing and availability is not currently available.

grade

FG
Fragrance grade
Halal
Kosher
natural

Quality Level

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117

vapor density

5.17 (vs air)

vapor pressure

1 mmHg ( 44 °C)

Assay

≥99%

autoignition temp.

914 °F

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

refractive index

n20/D 1.504 (lit.)

bp

212 °C (lit.)

mp

−34 °C (lit.)

density

1.045 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

greener alternative category

Organoleptic

fruity; floral

SMILES string

CCOC(=O)c1ccccc1

InChI

1S/C9H10O2/c1-2-11-9(10)8-6-4-3-5-7-8/h3-7H,2H2,1H3

InChI key

MTZQAGJQAFMTAQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Natural occurrence: Feijoa, guava, plum, raspberry, rum, strawberry, Virginia tobacco.
We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product is a Biobased products, showing key improvements in Green Chemistry Principles “Less Hazardous Chemical Syntheses” and “Use of Renewable Feedstock”.

Application


  • GC-MS Silylation Derivative Method to Characterise Black BIC Ballpoint 2-Phenoxyethanol Ratio Evaporation Profile-A Contribution to Ink Ageing Estimation.: This study uses GC-MS to analyze the evaporation profile of inks containing ethyl benzoate, aiding forensic science in the estimation of ink age on documents (Leal et al., 2023).

Biochem/physiol Actions

Taste at 10 ppm

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

190.4 °F - closed cup

Flash Point(C)

88 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organoleptic Characteristics of Flavor Materials
Mosciano, G.
Perfumer & Flavorist, 28, 102-102 (2003)
Daniel I Perez et al.
Bioorganic & medicinal chemistry, 17(19), 6914-6925 (2009-09-15)
Thienylhalomethylketones, whose chemical, biological, and pharmaceutical data are here reported, are the first irreversible inhibitors of GSK-3beta described to date. Their inhibitory activity is likely related to the cysteine residue present in the ATP-binding site, which is proposed as a
Alexandra Vinagre et al.
The journal of adhesive dentistry, 17(2), 107-116 (2015-04-29)
This study compared the microtensile bond strengths (μTBS) of two etch-and-rinse (ER) (OptiBond FL [OBFL]; Prime & Bond NT [PBNT]) and three self-etching (SE) (Clearfil SE Bond [CSEB]; Xeno III [XIII]; Xeno V+ [XV+]) adhesives systems to bur-prepared human enamel
Katarzyna Niedojadło et al.
Plant cell reports, 34(12), 2201-2215 (2015-09-12)
Calreticulin expression is upregulated during sexual reproduction of Hyacinthus orientalis, and the protein is localized both in the cytoplasm and a highly specialized cell wall within the female gametophyte. Several evidences indicate calreticulin (CRT) as an important calcium (Ca(2+))-binding protein
Suvi-Katriina Ruokonen et al.
Journal of chromatography. A, 1405, 178-187 (2015-06-15)
The effect of three phosphonium and imidazolium ionic liquids (ILs) on the interaction between liposomes and common pharmaceuticals found in wastewaters was studied. The liposomes comprised zwitterionic phosphatidyl choline and negatively charged phosphatidyl glycerol. A set of common cationic, anionic

Questions

  1. How much is the content of the sample pack? 50mg, 100mg, 500mg etc.

    1 answer
    1. The SAMPLE-K quantities vary from product to product and lot to lot. There is no established quantity assigned. These are typically gram quantities intended for use in formulation evaluation, however some more expensive compounds may contain smaller amounts.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service