Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W289302

Sigma-Aldrich

3-Phenylpropyl isobutyrate

≥98%, FG

Synonym(s):

3-Phenylpropyl 2-methylpropanoate, Hydrocinnamyl isobutyrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCO2(CH2)3C6H5
CAS Number:
Molecular Weight:
206.28
FEMA Number:
2893
EC Number:
Council of Europe no.:
303
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.428
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.515

Assay

≥98%

refractive index

n20/D 1.486 (lit.)

bp

282 °C (lit.)

density

0.979 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

balsam; fruity; sweet

SMILES string

CC(C)C(=O)OCCCc1ccccc1

InChI

1S/C13H18O2/c1-11(2)13(14)15-10-6-9-12-7-4-3-5-8-12/h3-5,7-8,11H,6,9-10H2,1-2H3

InChI key

VBTAKMZSMFMLGT-UHFFFAOYSA-N

Related Categories

General description

3-Phenylpropyl isobutyrate belongs to the cinnamyl phenylpropyl group of fragrance materials. It is the key volatile constituent of Roman camomile essential oil.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fragrance material review on 3-phenylpropyl isobutyrate.
Bhatia SP, et al.
Food And Chemical Toxicology, 49, S242-S245 (2011)
A toxicologic and dermatologic assessment of cinnamyl phenylpropyl materials when used as fragrance ingredients
Belsito D, et al.
Food And Chemical Toxicology, 49, S256-S267 (2011)
A toxicological and dermatological assessment of aryl alkyl alcohol simple acid ester derivatives when used as fragrance ingredients.
Belsito D, et al.
Food And Chemical Toxicology, 50(2), 2-2 (2012)
The Occurrence of 3-Phenylpropyl Isobutyrate in Roman Camomile Oil.
Bassols F & Thomas AF.
J. Essent. Oil Res., 3(5), 309-312 (1991)
The FEMA GRAS assessment of cinnamyl derivatives used as flavor ingredients.
Adams TB, et al.
Food And Chemical Toxicology, 42(2), 157-185 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service