Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W230901

Sigma-Aldrich

Citronellol

≥95%, FCC, FG

Synonym(s):

3,7-dimethyl-6-octenol, Elenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHCH2CH2CH(CH3)CH2CH2OH
CAS Number:
Molecular Weight:
156.27
FEMA Number:
2309
EC Number:
Council of Europe no.:
59
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.011
NACRES:
NA.21
grade:
FG
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

vapor pressure

~0.02 mmHg ( 25 °C)

Assay

≥95%

refractive index

n20/D 1.456 (lit.)

bp

225 °C (lit.)

density

0.855 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

waxy; floral; rose

SMILES string

CC(CCO)CC\C=C(\C)C

InChI

1S/C10H20O/c1-9(2)5-4-6-10(3)7-8-11/h5,10-11H,4,6-8H2,1-3H3

InChI key

QMVPMAAFGQKVCJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Citronellol is a volatile monoterpenic primary alcohol mainly found in the essential oil of plants such as Pelargonium graveolens, Cymbopogon winterianus and Rosa damascena. It is also one of the glycosidically bound aroma compounds in ginger.

Application


  • A comprehensive assessment of the cholinergic-supporting and cognitive-enhancing effects of Rosa damascena Mill. (Damask rose) essential oil on scopolamine-induced amnestic rats.: The research highlights citronellol′s role in enhancing cognitive function and supporting cholinergic systems, indicating its potential use in developing treatments for cognitive disorders (Teralı et al., 2024).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

224.6 °F - closed cup

Flash Point(C)

107 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antimicrobial activity and chemical composition of some essential oils
Ando?an BC, et al.
Archives of Pharmacal Research, 25(6), 860-864 (2002)
Phythochemical screening and anticonvulsant activity of Cymbopogon winterianus Jowitt (Poaceae) leaf essential oil in rodents
Quintans-Junior LJ, et al.
Phytomedicine, 15(8), 619-624 (2008)
Glycosidically bound aroma compounds in ginger (Zingiber officinale Roscoe).
Wu P, et al.
Journal of Agricultural and Food Chemistry, 38(7), 1553-1555 (1990)
Antibacterial effect of some essential oils administered alone or in combination with Norfloxacin
Rosato A, et al.
Phytomedicine, 14(11), 727-732 (2007)
Renan G Brito et al.
Basic & clinical pharmacology & toxicology, 112(4), 215-221 (2012-10-06)
Citronellol (CT) is a monoterpenoid alcohol present in the essential oil of many medicinal plants, such as Cymbopogon citratus. We evaluated the antinociceptive effects of CT on orofacial nociception in mice and investigated the central pathway involved in the effect.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service