Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

W281506

Sigma-Aldrich

Oleic acid

greener alternative

natural, FCC

Synonym(s):

cis-9-Octadecenoic acid, Elainic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7CH=CH(CH2)7COOH
CAS Number:
Molecular Weight:
282.46
FEMA Number:
2815
Beilstein:
1726542
EC Number:
Council of Europe no.:
13
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
8.013
NACRES:
NA.21
grade:
Fragrance grade
Halal
Kosher
natural
food allergen:
no known allergens
Pricing and availability is not currently available.

grade

Fragrance grade
Halal
Kosher
natural

Quality Level

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
FCC
FDA 21 CFR 172.210
FDA 21 CFR 172.860
FDA 21 CFR 182.70
FDA 21 CFR 182.90

vapor pressure

1 mmHg ( 176 °C)

form

liquid

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.459 (lit.)

bp

194-195 °C/1.2 mmHg (lit.)

mp

13-14 °C (lit.)

density

0.89 g/mL at 25 °C (lit.)

cation traces

As: ≤3 ppm
Cd: ≤1 ppm
Hg: ≤1 ppm
heavy metals (as Pb): <0.10 ppm

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

greener alternative category

Organoleptic

fatty

SMILES string

CCCCCCCC\C=C/CCCCCCCC(O)=O

InChI

1S/C18H34O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h9-10H,2-8,11-17H2,1H3,(H,19,20)/b10-9-

InChI key

ZQPPMHVWECSIRJ-KTKRTIGZSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Oleic acid is a monounsaturated fatty acid mainly found in high quantity in olive oil[1], peanut oil[2], sunflower oil[3] and canola oil.[4]
We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product is a Biobased products, showing key improvements in Green Chemistry Principles “Less Hazardous Chemical Syntheses” and “Use of Renewable Feedstock”.

Application


  • Orientation-Dependent Photoconductivity of Quasi-2D Nanocrystal Self-Assemblies.: Investigating the photoconductivity properties of quasi-2D nanocrystals, this research demonstrates the influence of oleic acid in manipulating the orientation and enhancing the electronic properties of these nanomaterials (Ibrahem et al., 2024).

  • Sequential gentle hydration increases encapsulation in model protocells.: Research in this article shows how oleic acid can be used to improve the encapsulation efficiency in model protocells, offering insights into the origins of cellular life (Gehlbach et al., 2024).

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genetic control of high oleic acid content in sunflower oil.
Miller JF, et al.
Crop Science, 27(5), 923-926 (1987)
Comparison of the effect of canola oil and sunflower oil on plasma lipids and lipoproteins and on in vivo thromboxane A2 and prostacyclin production in healthy young men.
McDonald BE, et al. (
American Journal of Clinical Nutrition, 50(6), 1382-1388 (1989)
Comparison of oxidative stability of high-and normal-oleic peanut oils.
O?keefe SF, et al.
Journal of the American Oil Chemists' Society, 70(5), 489-492 (1993)
Olive-oil consumption and health: the possible role of antioxidants.
Owen RW, et al.
Lancet Oncology, 1(2), 107-112 (2000)
Joyce Wang et al.
BMC genomics, 15, 415-415 (2014-06-03)
Mycobacterium avium subsp. paratuberculosis (MAP) is an obligate intracellular pathogen that infects many ruminant species. The acquisition of foreign genes via horizontal gene transfer has been postulated to contribute to its pathogenesis, as these genetic elements are absent from its

Questions

  1. What is the average purity of oleic acid in this product? What are the main impurities? and how much linoleic acid it roughly contains?

    1 answer
    1. The oleic and linoleic concentrations vary somewhat from lot to lot. The product is tested for heavy metals, however no other potential contaminants are identified. Please see the link below to access a sample or lot specific Certificate:
      https://www.sigmaaldrich.com/product/aldrich/w281506#product-documentation

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service