Skip to Content
MilliporeSigma
All Photos(3)

Documents

W236012

Sigma-Aldrich

(+)-γ-Decalactone

greener alternative

natural, ≥97%, FG

Synonym(s):

(+)-gamma-Decalone

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2360
EC Number:
Council of Europe no.:
2230c
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
10.017
NACRES:
NA.21

grade

FG
Fragrance grade
Halal
Kosher
natural

Quality Level

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥97%

optical activity

[α]24/D +34°, neat

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.449

bp

281 °C

density

0.952 g/mL at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

greener alternative category

Organoleptic

creamy; fruity; peach; sweet

SMILES string

CCCCCC[C@@H]1CCC(=O)O1

InChI

1S/C10H18O2/c1-2-3-4-5-6-9-7-8-10(11)12-9/h9H,2-8H2,1H3/t9-/m1/s1

InChI key

IFYYFLINQYPWGJ-SECBINFHSA-N

General description

(+)-γ-Decalactone, the main volatile aroma compound found in peaches, can be used as a flavoring agent in the food industry due to its distinct peach odor. It is also found in strawberries and apricots.
We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product is a Biobased products, showing key improvements in Green Chemistry Principles “Less Hazardous Chemical Syntheses” and “Use of Renewable Feedstock”.

Application


  • Decoding the Effect of Running on Flavor Perception Changes during Consumption of Sports Drinks.: Pu et al. investigate how physical activity affects flavor perception, particularly focusing on sports drinks. The study includes (+)-γ-Decalactone as a compound of interest due to its impact on flavor modulation during and after exercise (Pu et al., 2024).

  • Analysis of volatile organic compounds in Korean-bred strawberries: insights for improving fruit flavor.: Jee et al. analyze volatile organic compounds in strawberries, identifying key compounds such as (+)-γ-Decalactone that contribute to fruit flavor. This research provides valuable insights for the enhancement of fruit taste in breeding programs (Jee et al., 2024).

  • (R)-(+)-γ-Decalactone is Conserved in North America as a Pheromone Component of Osmoderma eremicola (Coleoptera: Scarabaeidae) and a Kairomone of Elater abruptus (Coleoptera: Elateridae).: Steffek et al. examine the role of (+)-γ-Decalactone as a pheromone and kairomone in specific beetle species, highlighting its ecological and biochemical significance (Steffek et al., 2024).

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

276.8 °F - closed cup

Flash Point(C)

136 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aroma characterization of various apricot varieties using headspace?solid phase microextraction combined with gas chromatography?mass spectrometry and gas chromatography?olfactometry.
Guillot S, et al.
Food Chemistry, 96(1), 147-155 (2006)
Microbial production of flavors for the food industry. A case study on the production of gamma-decalactone, the key compound of peach flavor, by the yeasts Sporidiobolus sp
Dufosse L, et al.
Food Science and Biotechnology, 11(2), 192-202 (2002)
Gilles Feron et al.
Canadian journal of microbiology, 53(6), 738-749 (2007-08-02)
The sensitivity of Sporidiobolus ruinenii yeast to the use of reducing agents, reflected in changes in the oxidoreduction potential at pH 7 (Eh7) environment, ricinoleic acid methyl ester catabolism, gamma-decalactone synthesis, cofactor level, beta-oxidation activity, and ultrastructure of the cell
Analysis of strawberry flavour?discrimination of aroma types by quantification of volatile compounds.
Ulrich D, et al.
European Food Research and Technology, 205(3), 218-223 (1997)

Articles

Recent regulatory changes in the US and EU have put the focus on fragrance safety and ingredient transparency, including the MoCRA, to be implemented within the next two years. Manufacturers and suppliers must comply with these changes to ensure they are meeting consumer demand and industry expectations for safety.

Recent regulatory changes in the US and EU have put the focus on fragrance safety and ingredient transparency, including the MoCRA, to be implemented within the next two years. Manufacturers and suppliers must comply with these changes to ensure they are meeting consumer demand and industry expectations for safety.

Recent regulatory changes in the US and EU have put the focus on fragrance safety and ingredient transparency, including the MoCRA, to be implemented within the next two years. Manufacturers and suppliers must comply with these changes to ensure they are meeting consumer demand and industry expectations for safety.

Recent regulatory changes in the US and EU have put the focus on fragrance safety and ingredient transparency, including the MoCRA, to be implemented within the next two years. Manufacturers and suppliers must comply with these changes to ensure they are meeting consumer demand and industry expectations for safety.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service