Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

W309109

Sigma-Aldrich

γ-Undecalactone

≥98%, FCC, FG

Synonym(s):

Undecanoic γ-lactone, Aldehyde C14

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H20O2
CAS Number:
Molecular Weight:
184.28
FEMA Number:
3091
EC Number:
Council of Europe no.:
179
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
10.002
NACRES:
NA.21
Organoleptic:
coconut; creamy; fatty; fruity; peach
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
Agency:
follows IFRA guidelines
meets purity specifications of JECFA
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 172.515

Assay

≥98%

refractive index

n20/D 1.45 (lit.)

bp

164-166 °C/13 mmHg (lit.)

density

0.949 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

coconut; creamy; fatty; fruity; peach

SMILES string

CCCCCCCC1CCC(=O)O1

InChI

1S/C11H20O2/c1-2-3-4-5-6-7-10-8-9-11(12)13-10/h10H,2-9H2,1H3

InChI key

PHXATPHONSXBIL-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)

Looking for similar products? Visit Product Comparison Guide

General description

γ-Undecalactone is a volatile flavor constituent found in low-fat ice cream.

Application


  • Investigation of Lactone Chiral Enantiomers and Their Contribution to the Aroma of Longjing Tea by Odor Activity Value and S-Curve.: This study explores the chiral enantiomers of lactones, including γ-Undecalactone, and their impact on the aroma profile of Longjing tea, using odor activity value and S-curve methods to quantify their contributions (Zhu et al., 2023).

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

293.0 °F - closed cup

Flash Point(C)

145 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Total phenolic compounds, antioxidant activity and nutritional values of sugar-free and reduced-fat milk-based ice cream enriched with selected herb ingredients.
Limsuwan T.
Asia-Pacific Journal of Science and Technology , 19(4), 515-525 (2017)
Laura E Korhonen et al.
Journal of medicinal chemistry, 48(11), 3808-3815 (2005-05-27)
The purpose of this study was to determine the cytochrome P450 1A2 (CYP1A2) inhibition potencies of structurally diverse compounds to create a comprehensive three-dimensional quantitative structure-activity relationship (3D-QSAR) model of CYP1A2 inhibitors and to use this model to predict the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service