Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

W236128

Sigma-Aldrich

δ-Decalactone

greener alternative

natural, ≥98%, FCC, FG

Synonym(s):

(±)-5-Decanolide, δ-Decanolactone, (±)-δ-Pentyl-δ-valerolactone, (±)-6-Pentyltetrahydro-2H-pyran-2-one, 5-Hydroxydecanoic acid δ-lactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O2
CAS Number:
Molecular Weight:
170.25
FEMA Number:
2361
Beilstein:
117520
EC Number:
Council of Europe no.:
621
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
10.007
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
natural
food allergen:
no known allergens

grade

FG
Fragrance grade
Halal
Kosher
natural

Quality Level

300
400

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117

Assay

≥98%

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.458 (lit.)

bp

117-120 °C/0.02 mmHg (lit.)

mp

−27 °C (lit.)

density

0.954 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

greener alternative category

Organoleptic

coconut; creamy; fatty; milk; peach; sweet

SMILES string

CCCCCC1CCCC(=O)O1

InChI

1S/C10H18O2/c1-2-3-4-6-9-7-5-8-10(11)12-9/h9H,2-8H2,1H3

InChI key

GHBSPIPJMLAMEP-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)

Looking for similar products? Visit Product Comparison Guide

General description

δ-Decalactone is the main flavor compound found in nonfat dry milk, evaporated and dried whole milk. It is reported to be responsible for the coconut-like off-flavor in fat-containing dairy products.
Natural occurrence: Sherry, white wine, mango, cassava, loganberry and fresh plum.
We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product is a Biobased products, showing key improvements in Green Chemistry Principles “Less Hazardous Chemical Syntheses” and “Use of Renewable Feedstock”.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aroma-active components of nonfat dry milk
Karagul-Yuceer Y, et al.
Journal of Agricultural and Food Chemistry, 49(6), 2948-2953 (2001)
Gas Chromatographic Analysis of Flavor in Processed Milks1, 2.
Patton S.
The Journal of Dairy Research, 44(2), 207-214 (1961)
Mosciano, G.
Perfumer & Flavorist, 21, 51-51 (1996)
The Coconut-Like Flavor Defect of Milk Fat. II. Demonstration of ?-Decalactone in Dried Cream, Dry Whole Milk, and Evaporated Milk1.
Keeney PG & Patton S.
The Journal of Dairy Research, 39(8), 1114-1119 (1956)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service