Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SYNC195262C

Synthware chromatography column with spherical joints and coarse fritted disc

joint: ST/NS 35/20, I.D. × L 20.0 mm × 254 mm

Synonym(s):

Synthware C195262C, glass chromatograpy column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.51

Joint

joint: ST/NS35/20

grade

for chromatography

packaging

pkg of 1 ea

manufacturer/tradename

Synthware C195262C

I.D. × L

20.0 mm × 254 mm

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

With a coarse porosity fritted disc to support packing material. All Synthware columns are heavy wall designed to increase safety.

Legal Information

Synthware is a trademark of Kemtech America Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service