Key Documents
Z163961
Sigma-Aldrich
Gravity column
Short
Synonym(s):
Gravity Filter
About This Item
UNSPSC Code:
23151816
NACRES:
NB.51
Pricing and availability is not currently available.
Joint
joint ST/NS24/40
L × I.D. × O.D.
33 cm × 24 mm × 28 mm
Related Categories
General description
Gravity column can prevent special moment frame structures from collapsing.[1]
Features and Benefits
  • Provided with a 4mm polytetrafluoroethylene (PTFE) stopcock
  • Has no frit and uses a glass-wool plug and sand
  • Use along with coarse silica gel (e.g., Cat. No. 60744) for fast and general-purpose purification.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
The influence of gravity column continuity on the seismic performance of special steel moment frame structures
Flores F, et al.
ACC Journal, 118, 217-230 (2016)