MilliporeSigma
SYNC195173
Synthware chromatography column with with spherical joint and PTFE stopcock
joint: ST/NS 35/20, I.D. × L 13.4 mm × 305 mm
Synonym(s):
Synthware C195173, glass chromatograpy column
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.51
UNSPSC Code:
41121800
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
joint
joint: ST/NS35/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware C195173
I.D. × L
13.4 mm × 305 mm
suitability
suitable for chromatography
General description
Spherical joint used with pinch clamp makes quick and tight connection under pressure for flash chromatography. All Synthware columns are heavy wall designed to increase safety.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)