Key Documents
Z223360
Sigma-Aldrich
Aldrich® three-way adapter with thermometer joint
joint: ST/NS 24/40
Synonym(s):
adapter laboratory, glass adapter, glass adapters
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
Pricing and availability is not currently available.
Joint
thermometer joint ST/NS10/30
joint: ST/NS24/40
General description
Side-arm is at 75° angle from bottom joint. Distance from joint to side-arm is 75mm.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)