Key Documents
SYNA288250
Synthware anti-splash adapter
250 mL, top joint: ST/NS 29/42, bottom joint: ST/NS 29/42
Synonym(s):
Synthware A288250, splash-guard adapter
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
Joint
bottom joint: ST/NS29/42
joint: ST/NS top29/42
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A288250
capacity
250 mL
General description
This design prevents possible contamination from rotary evaporator.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)