Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z549290

Sigma-Aldrich

Aldrich® antisplash splash-guard adapter with return hole

female joint: ST/NS 29/32, male joint: ST/NS 29/32, capacity 100 mL

Synonym(s):

Antisplash Adapter, Splash Guard with Return Hole, Splash-Guard Adapter

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121815
NACRES:
NB.35

Joint

female joint: ST/NS29/32
male joint: ST/NS29/32

capacity

100 mL

Looking for similar products? Visit Product Comparison Guide

General description

These traps greatly reduce carry-over from foaming or bumping. Anything that may climb can return back down through the return hole at the bottom of this adapter.

Application

Aldrich antisplash splash-guard adapter with return hole may be used between the vapor tube and rotary evaporator flask to prevent solvent from being drawn into the condenser in case of "bumping". A drain hole at the base of the inner tube allows solvent to drain back into the evaporation flask.

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service