Skip to Content
MilliporeSigma
All Photos(1)

Documents

SYNC182402

Synthware chromatography reservoir

capacity 250 mL, joint: ST/NS 24/40

Synonym(s):

Synthware C182402, Glass chromatography reservoir

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.51

material

colorless
glass

Joint

joint: ST/NS24/40

grade

for chromatography

sterility

non-sterile

packaging

pkg of 1 ea

manufacturer/tradename

Synthware C182402

capacity

250 mL

Looking for similar products? Visit Product Comparison Guide

General description

For flash chromatography.

Legal Information

Synthware is a trademark of Kemtech America Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service