Key Documents
SYNA282250
Synthware anti-splash adapter
250 mL, top joint: ST/NS 29/42, bottom joint: ST/NS 24/40
Synonym(s):
Synthware A282250, rotary evaporator bump trap, bump trap
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
Joint
bottom joint: ST/NS24/40
joint: ST/NS top29/42
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A282250
capacity
250 mL
Related Categories
General description
This design prevents possible contamination from rotary evaporator.
Legal Information
Synthware is a trademark of Kemtech America Inc.