Skip to Content
MilliporeSigma
All Photos(1)

Documents

900644

Sigma-Aldrich

Acetonitrile

99.8%, ZerO2®, anhydrous

Synonym(s):

Acetonitrile ZerO2®, Degassed and low oxygen acetonitrile, ACN, Cyanomethane, Ethyl nitrile, Methyl cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CN
CAS Number:
Molecular Weight:
41.05
Beilstein:
741857
EC Number:
MDL number:
UNSPSC Code:
12191502
NACRES:
NA.21

product name

Acetonitrile, anhydrous, ZerO2®, 99.8%

grade

ZerO2®

Quality Level

vapor density

1.41 (vs air)

vapor pressure

72.8 mmHg ( 20 °C)

Assay

99.8%

form

liquid

autoignition temp.

973 °F

quality

anhydrous

expl. lim.

16 %

impurities

≤0.001% water
water

refractive index

n20/D 1.344 (lit.)

bp

81-82 °C (lit.)

mp

−45 °C (lit.)

density

0.786 g/mL at 25 °C (lit.)

format

neat

SMILES string

CC#N

InChI

1S/C2H3N/c1-2-3/h1H3

InChI key

WEVYAHXRMPXWCK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

ZerO2 products are rigorously degassed with highly pure inert gas providing solvents and solutions (anhydrous if specified) with very low residual oxygen content.

Legal Information

ZerO2 is a registered trademark of Sigma-Aldrich Co. LLC

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service