Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

D181803

Sigma-Aldrich

2,6-Dimethylpyrazine

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8N2
CAS Number:
Molecular Weight:
108.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

bp

154 °C (lit.)

mp

35-40 °C (lit.)

SMILES string

Cc1cncc(C)n1

InChI

1S/C6H8N2/c1-5-3-7-4-6(2)8-5/h3-4H,1-2H3

InChI key

HJFZAYHYIWGLNL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

127.4 °F - closed cup

Flash Point(C)

53 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nobuo Ochiai et al.
Journal of chromatography. A, 1421, 103-113 (2015-08-16)
An extension of multi-volatile method (MVM) technology using the combination of a standard dynamic headspace (DHS) configuration, and a modified DHS configuration incorporating an additional vacuum module, was developed for milliliter injection volume of aqueous sample with full sample evaporation.
Gustavo Luis Leonardo Scalone et al.
Journal of agricultural and food chemistry, 63(22), 5364-5372 (2015-05-15)
Pyrazines are specific Maillard reaction compounds known to contribute to the unique aroma of many products. Most studies concerning the generation of pyrazines in the Maillard reaction have focused on amino acids, while little information is available on the impact
Enbo Xu et al.
Journal of food science, 80(7), C1476-C1489 (2015-06-20)
Enzymatic extrusion, instead of traditional steam cooking, to treat rice is an efficient and alternative pretreatment for Chinese rice wine fermentation. In order to determine the formation of volatiles in enzymatic extrusion-processed rice wine (EE), and to confirm its characteristic

Protocols

Separation of various pyrazines and sulfur compounds including carbon disulfide and dimethyl disulfide.

Separation of various pyrazines and sulfur compounds including carbon disulfide and dimethyl disulfide.

Separation of various pyrazines and sulfur compounds including carbon disulfide and dimethyl disulfide.

Separation of various pyrazines and sulfur compounds including carbon disulfide and dimethyl disulfide.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service