Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CLS431750

Corning® cell strainer

pore size 40 μm, blue, sterile, pkg of (individually wrapped), pack of 50 ea

Synonym(s):

Corning cell strainer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104932
NACRES:
NB.24

material

Nylon
blue

sterility

sterile

packaging

pack of 50 ea
pkg of (individually wrapped)

manufacturer/tradename

Corning 431750

pore size

40 μm

Looking for similar products? Visit Product Comparison Guide

General description

Corning® 40µm Cell Strainer is a sterile, easy-to-use device for rapidly isolating primary cells to consistently obtain a uniform single-cell suspension from tissues. Applications include stem cells and primary cells. Individually packaged, 50/case.

Application

  • Faster, easier alternative to gauze filtration
  • Ideal for preparation of flow cytometry samples, including:
    Single cell suspensions of blood cells from marrow, pancreas, thymus, tonsil, and lymph nodes
    Stem cells, tissue-derived cells, and cancer cells
    Preparation of specimens for primary cell cultures and immunogens
    Preparation of freezing stocks
    Filtering agglutinative proteins produced in inactivation serum

Features and Benefits

  • Strong nylon mesh with 40- micron pores for optimal performance in a variety of applications
  • Evenly spaced mesh pores providing consistent and reliable results
  • Conveniently accessible in individual packaging
  • Extended lip on strainer enables aseptic handling with forceps
  • Molded color-coded polypropylene frame with tab enables easy handling and identification
  • Fits all major brands of 50mL Conical Tube
  • Disposable, easy-to-use, inexpensive, maintains sample integrity
  • Sterilized by gamma irradiation, noncytotoxic

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel Ley et al.
Metabolic engineering, 56, 120-129 (2019-09-19)
Chinese hamster ovary (CHO) cells are the preferred host for producing biopharmaceuticals. Amino acids are biologically important precursors for CHO metabolism; they serve as building blocks for proteogenesis, including synthesis of biomass and recombinant proteins, and are utilized for growth
Jiaxing Li et al.
Frontiers in cell and developmental biology, 9, 598364-598364 (2021-02-16)
Decidualization is driven by differentiation of human endometrial stromal cells (ESCs), and is a prerequisite for successful implantation and establishment of pregnancy. The critical role of impaired decidualization in women suffered recurrent implantation failure (RIF) has been established, while the
Joseph Park et al.
Nature medicine, 27(1), 66-72 (2021-01-13)
The clinical impact of rare loss-of-function variants has yet to be determined for most genes. Integration of DNA sequencing data with electronic health records (EHRs) could enhance our understanding of the contribution of rare genetic variation to human disease1. By
Ziqiang Wang et al.
Cellular and molecular life sciences : CMLS, 77(17), 3441-3452 (2019-12-16)
The elevated expression of immune checkpoints by the tumor microenvironment is associated with poor prognosis in several cancers due to the exhaustion of tumor-infiltrating lymphocytes (TILs), and the effective suppression of the expression of these genes is key to reversing
Joanna Kalucka et al.
Cell, 180(4), 764-779 (2020-02-16)
The heterogeneity of endothelial cells (ECs) across tissues remains incompletely inventoried. We constructed an atlas of >32,000 single-EC transcriptomes from 11 mouse tissues and identified 78 EC subclusters, including Aqp7+ intestinal capillaries and angiogenic ECs in healthy tissues. ECs from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service