Skip to Content
MilliporeSigma
All Photos(1)

Documents

199419

Sigma-Aldrich

2,3,5-Trimethylpyrazine

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10N2
CAS Number:
Molecular Weight:
122.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.5040 (lit.)

bp

171-172 °C (lit.)

density

0.979 g/mL at 25 °C (lit.)

SMILES string

Cc1cnc(C)c(C)n1

InChI

1S/C7H10N2/c1-5-4-8-6(2)7(3)9-5/h4H,1-3H3

InChI key

IAEGWXHKWJGQAZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,3,5-Trimethylpyrazine is present in brown cheese.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

129.2 °F - closed cup

Flash Point(C)

54 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R D Divine et al.
Journal of dairy science, 95(5), 2347-2354 (2012-05-01)
Brown pigmentation can occasionally form in Parmesan cheese during the ripening process, creating an unappealing appearance and associated off-flavors. The browning reactions proceed at refrigerated temperatures and in the relative absence of reducing sugar, contrary to typical Maillard browning. Residual
Deng-Rong Xiao et al.
Scientific reports, 5, 17120-17120 (2015-11-27)
The aroma attributes of sulfurous, mushroom and earthy are the most important characteristics of the aroma of Tuber melanosporum. However, these three aroma attributes are absent in the T. melanosporum fermentation system. To improve the quality of the aroma, repeated
JianCai Zhu et al.
Journal of agricultural and food chemistry, 63(34), 7499-7510 (2015-08-11)
The aroma profile of oolong tea infusions (Dongdingwulong, DDWL; Tieguanyin, TGY; Dahongpao, DHP) were investigated in this study. Gas chromatography-olfactometry (GC-O) with the method of aroma intensity (AI) was employed to investigate the aroma-active compounds in tea infusions. The results

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service