Key Documents
BR452010
BRAND® disposable petri dish with lid
L × W 55 mm × 16 mm, vented
Synonym(s):
Dishes, Petri Dishes, Plastics
About This Item
UNSPSC Code:
41161502
NACRES:
NB.14
material
polystyrene
feature
vented
manufacturer/tradename
BRAND 452010
L × W
55 mm × 16 mm
Related Categories
General description
Made from high-quality polystyrene (PS), these transparent dishes lids provide optimal visibility for observing cultures. Dishes and lids are robotically assembled to significantly reduce the risk of contamination, ensuring the integrity of your samples.
Features and Benefits
  • BRAND® disposable petri dish is manufactured from excellent quality polystyrene for sample clarity and visibility.
  • The transparent dishes and lids are assembled robotically to reduce the risk of contamination.
  • Able to withstand temperature up to 80 °C.
  • Fully stackable.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)