Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

936448

Sigma-Aldrich

Fmoc-Gly-Gly-Gly-OH

≥95%

Synonym(s):

2-[2-[2-([[(9H-Fluoren-9-yl)methoxy]carbonyl]amino)acetamido]acetamido]acetic acid, 2-[[2-[[2-(9H-Fluoren-9-ylmethoxycarbonylamino)acetyl]amino]acetyl]amino]acetic acid, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]glycylglycylglycine, Glycine, N-[(9H-fluoren-9-ylmethoxy)carbonyl]glycylglycyl-, Glycine, N-[N-[N-[(9H-fluoren-9-ylmethoxy)carbonyl]glycyl]glycyl]-

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H21N3O6
CAS Number:
Molecular Weight:
411.41
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.22

Quality Level

Assay

≥95%

form

powder or crystals

color

white to off-white

storage temp.

2-8°C

SMILES string

O=C(OCC1C=2C=CC=CC2C=3C=CC=CC31)NCC(=O)NCC(=O)NCC(=O)O

InChI

InChI= 1S/C21H21N3O6/c25-18(23-11-20(27)28)9-22-19(26)10-24-21(29)30-12-17-15-7-3-1-5-13(15)14-6-2-4-8-16(14)17/h1-8,17H,9-12H2,(H,22,26)(H,23,25)(H,24,29)(H,27,28)

Application

Fmoc-Gly-Gly-Gly-OH is a lysosomally cleavable linker used in construction of antibody-drug conjugates (ADC & Bioconjugation).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Strategies and challenges for the next generation of antibody-drug conjugates
Beck A, et al.
Nature Reviews Drug Discovery, 16, 315-337 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service