Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

936413

Sigma-Aldrich

Fmoc-Val-Cit-OH

≥95%

Synonym(s):

(S)-2-((S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-methylbutanamido)-5-ureidopentanoicacid, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-valyl-N5-(aminocarbonyl)-L-ornithine (ACI), L-Ornithine, N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-valyl-N5-(aminocarbonyl)-, L-Ornithine, N5-(aminocarbonyl)-N2-[N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-valyl]- (ZCI)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H32N4O6
CAS Number:
Molecular Weight:
496.56
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.22

Quality Level

Assay

≥95%

form

powder or crystals

color

white to beige

storage temp.

2-8°C

SMILES string

C(OC(N[C@H](C(N[C@@H](CCCNC(N)=O)C(O)=O)=O)C(C)C)=O)C1C=2C(C=3C1=CC=CC3)=CC=CC2

InChI

InChI=1S/C26H32N4O6/c1-15(2)22(23(31)29-21(24(32)33)12-7-13-28-25(27)34)30-26(35)36-14-20-18-10-5-3-8-16(18)17-9-4-6-11-19(17)20/h3-6,8-11,15,20-22H,7,12-14H2,1-2H3,(H,29,31)(H,30,35)(H,32,33)(H3,27,28,34)/t21-,22-/m0/s1

InChI key

ZJLNQOIFTYLCHC-VXKWHMMOSA-N

Application

This is a cathepsin cleavable linker used in bioconjugation, specifically in the construction of antibody drug conjugates (ADC & Bioconjugation).

Features and Benefits

Fmoc-Val-Cit-OH is a cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADC). The Val-Cit is specifically cleaved by cathepsin B.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cryptophycin-55/52 based antibody-drug conjugates: Synthesis, efficacy, and mode of action studies
Lai Q, et al.
European Journal of Medicinal Chemistry, 199, 112364-112364 (2020)
EGFR Targeted Cetuximab-Valine-Citrulline (vc)-Doxorubicin Immunoconjugates- Loaded Bovine Serum Albumin (BSA) Nanoparticles for Colorectal Tumor Therapy
Ye Z, et al.
International journal of nanomedicine, 26, 2443-2459 (2021)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service