Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

47625

Sigma-Aldrich

Fmoc-Glu(OtBu)-OH

≥98.0% (HPLC), for peptide synthesis

Synonym(s):

Fmoc-L-glutamic acid 5-tert-butyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H27NO6
CAS Number:
Molecular Weight:
425.47
Beilstein:
3636375
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

Product Name

Fmoc-Glu(OtBu)-OH, ≥98.0% (HPLC)

Assay

≥98.0% (HPLC)

form

powder

optical activity

[α]20/D −5.0±2.0°, c = 1% in acetic acid: water (4:1)

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

impurities

~4% water

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)CC[C@H](NC(=O)OCC1c2ccccc2-c3ccccc13)C(O)=O

InChI

1S/C24H27NO6/c1-24(2,3)31-21(26)13-12-20(22(27)28)25-23(29)30-14-19-17-10-6-4-8-15(17)16-9-5-7-11-18(16)19/h4-11,19-20H,12-14H2,1-3H3,(H,25,29)(H,27,28)/t20-/m0/s1

InChI key

OTKXCALUHMPIGM-FQEVSTJZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Fmoc-Glu(OtBu)-OH is widely used as a building block in peptide synthesis for the protection of amine groups.

Application

Fmoc-Glu(OtBu)-OH (Fmoc-L-glutamic acid 5-tert-butyl ester) can be employed as a:
  • Ligand in the synthesis of cis-substituted cyclopropane carboxylic acids via C-H activation of cyclopropane carboxamides using Pd catalyst.
  • Linker in the preparation of multi-small molecule-conjugated PTX (paclitaxel) derivatives.

It can be also used as a building block in the preparation of stapled α-helical peptides , and peptide C-terminal thioesters .

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solid phase synthesis of peptide C-terminal thioesters by Fmoc/t-Bu chemistry
Ingenito R, et al.
Journal of the American Chemical Society, 121(49), 11369-11374 (1999)
Pd (II)-catalyzed enantioselective C-H activation of cyclopropanes
Wasa M, et al.
Journal of the American Chemical Society, 133(49), 19598-19601 (2011)
Stapled α-helical peptide drug development: A potent dual inhibitor of MDM2 and MDMX for p53-dependent cancer therapy
Chang YS, et al.
Proceedings of the National Academy of Sciences of the USA, 110(36), E3445-E3454 (2013)
Synthesis of peptides from alpha-and beta-tubulin containing glutamic acid side-chain linked oligo-glu with defined length
Tegge W, et al.
International Journal of Peptides, 2010 (2010)
Multi-small molecule conjugations as new targeted delivery carriers for tumor therapy
Shan L, et al.
International journal of nanomedicine, 10(49), 5571-5571 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service