Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MSQC20

Sigma-Aldrich

SILuLite SigmaMAb Bevacizumab Monoclonal Antibody

recombinant, expressed in CHO cells

Synonym(s):

Mass spectrometry standard, Bevacizumab

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

recombinant

expressed in CHO cells

Assay

≥90% (SDS-PAGE)

form

solid

suitability

suitable for mass spectrometry

shipped in

wet ice

storage temp.

−20°C

Related Categories

General description

SigmaMAb Bevacizumab (MSQC20) was designed to be a standard for the analysis of Bevacizumab monoclonal antibodies using mass spectrometry. An isotopically labeled version is available as SILuMab Bevacizumab (MSQC21).

SigmaMAb Bevacizumab is for R&D use only. Not for drug, household, or other uses.

Application

SILu Lite SigmaMAb Bevacizumab Monoclonal Antibody has been used as a proangiogenic factor to study its role in the angiogenesis process by Raman spectroscopy.

Biochem/physiol Actions

Bevacizumab serves as an antitumor agent. Studies show that chemotherapy using bevacizumab improves the condition of metastatic colorectal cancer and renal cell cancer patients.

Sequence

SigmaMab Bevacizumab Heavy Chain:
EVQLVESGGGLVQPGGSLRLSCAASGYTFTNYGMNWVRQAPGKGLEWVGWINTYTGEPTYAADFKRRFTFSLDTSKSTAYLQMNSLRAEDTAVYYCAKYPHYYGSSHWYFDVWGQGTLVTVSSASTKGPSVFPLAPSSKSTSGGTAALGCLVKDYFPEPVTVSWNSGALTSGVHTFPAVLQSSGLYSLSSVVTVPSSSLGTQTYICNVNHKPSNTKVDKKVEPKSCDKTHTCPPCPAPELLGGPSVFLFPPKPKDTLMISRTPEVTCVVVDVSHEDPEVKFNWYVDGVEVHNAKTKPREEQYNSTYRVVSVLTVLHQDWLNGKEYKCKVSNKALPAPIEKTISKAKGQPREPQVYTLPPSREEMTKNQVSLTCLVKGFYPSDIAVEWESNGQPENNYKTTPPVLDSDGSFFLYSKLTVDKSRWQQGNVFSCSVMHEALHNHYTQKSLSLSPG

SigmaMab Bevacizumab Light Chain:
DIQMTQSPSSLSASVGDRVTITCSASQDISNYLNWYQQKPGKAPKVLIYFTSSLHSGVPSRFSGSGSGTDFTLTISSLQPEDFATYYCQQYSTVPWTFGQGTKVEIKRTVAAPSVFIFPPSDEQLKSGTASVVCLLNNFYPREAKVQWKVDNALQSGNSQESVTEQDSKDSTYSLSSTLTLSKADYEKHKVYACEVTHQGLSSPVTKSFNRGEC

Legal Information

SILu is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service