Skip to Content
MilliporeSigma
All Photos(1)

Documents

933201

Sigma-Aldrich

SEM Sample Holder

TEM Grid Holder on Pin

Synonym(s):

multiple holder on pin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41111761
NACRES:
NB.35

General description

EMS techware
The overall diameter of the holder is 0.7" (18 mm) with a ⅛" diameter pin (3.2 mm) which is 0.6" (15 mm) long. The grid locations are all numbered.

Application

This EMS new release allows for the holding of up to 4 grids. Made from aluminum with a brass screw this holder allows you to image and analyze specimens on TEM Grids in the SEM.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service