Documents
TEM-CF313-25
Sigma-Aldrich
C-flat Holey Carbon Grids (CF-1.2/1.3) for TEM
Copper Grids, size 300 mesh, pack of 25
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
packaging
pack of 25
size
300 mesh
General description
C-flat Holey Carbon Grids for TEM, Copper Grids, 300 Mesh, 1.2 micron hole, 1.3 micron spacing, 25/pk