Skip to Content
MilliporeSigma
All Photos(1)

Documents

931500

Sigma-Aldrich

TEM Grids

Carbon Film Supported Copper Grid, Standard Thickness, grid size 400 mesh, box of 50

Synonym(s):

TEM grids

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41111761
NACRES:
MA.24

product name

Carbon Film Supported Copper Grid, Standard Thickness, grid size 400 mesh, box of 50

material

carbon film
copper grid

packaging

box of 50

film size

5-6 nm

grid size

400 mesh

Looking for similar products? Visit Product Comparison Guide

General description

TEM grids supported by carbon film only. A 5-6 nm thin film of pure carbon deposited on one side of the standard square mesh grid.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service