Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

901197

Sigma-Aldrich

(t-Bu)2PhP Pd G3

95%

Synonym(s):

Methanesulfonato (di-tert-butyl) phenylphosphino (2′-amino-1,1′-biphenyl-2-yl) palladium(II), PPh(t-Bu)2-Pd-G3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H36NO3PPdS
CAS Number:
Molecular Weight:
592.04
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.22

Quality Level

Assay

95%

form

solid

feature

generation 3

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

mp

163-169 °C (decomp.)

SMILES string

CC(C)(C)P(C(C)(C)C)C1=CC=CC=C1.NC2=C(C3=C([Pd]OS(C)(=O)=O)C=CC=C3)C=CC=C2

InChI

1S/C14H23P.C12H10N.CH4O3S.Pd/c1-13(2,3)15(14(4,5)6)12-10-8-7-9-11-12;13-12-9-5-4-8-11(12)10-6-2-1-3-7-10;1-5(2,3)4;/h7-11H,1-6H3;1-6,8-9H,13H2;1H3,(H,2,3,4);/q;;;+1/p-1

InChI key

IQWMOOOMAPPAMZ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Other Notes

Third-generation Buchwald precatalyst.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service