Skip to Content
MilliporeSigma
All Photos(2)

Documents

804851

Sigma-Aldrich

Mesyl[(tri-t-butylphosphine)-2-(2-aminobiphenyl)]palladium(II)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H40NO3PPdS
CAS Number:
Molecular Weight:
572.05
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

powder

Quality Level

feature

generation 3

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: Cross Couplings

mp

157-162 °C (w/ decomp)

functional group

phosphine

SMILES string

NC1=C(C2=C([Pd]OS(C)(=O)=O)C=CC=C2)C=CC=C1.CC(P(C(C)(C)C)C(C)(C)C)(C)C

InChI

1S/C12H10N.C12H27P.CH4O3S.Pd/c13-12-9-5-4-8-11(12)10-6-2-1-3-7-10;1-10(2,3)13(11(4,5)6)12(7,8)9;1-5(2,3)4;/h1-6,8-9H,13H2;1-9H3;1H3,(H,2,3,4);/q;;;+1/p-1

InChI key

ZLJAFHQENIQAOV-UHFFFAOYSA-M

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

G3 and G4 Buchwald palladium precatalysts are the newest air, moisture, and thermally stable crossing-coupling complexes used in bond formation for their versatility and high reactivity.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service