Skip to Content
MilliporeSigma
All Photos(6)

Documents

756482

Sigma-Aldrich

P(t-Bu)3 Pd G2

Synonym(s):

Chloro[(tri-tert-butylphosphine)-2-(2-aminobiphenyl)] palladium(II)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H37ClNPPd
CAS Number:
Molecular Weight:
512.40
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

solid

Quality Level

feature

generation 2

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: Cross Couplings

mp

167-170 °C (decomposition)

functional group

phosphine

SMILES string

NC1=C(C2=CC=CC=C2[Pd]Cl)C=CC=C1.CC(C)(C)P(C(C)(C)C)C(C)(C)C

InChI

1S/C12H10N.C12H27P.ClH.Pd/c13-12-9-5-4-8-11(12)10-6-2-1-3-7-10;1-10(2,3)13(11(4,5)6)12(7,8)9;;/h1-6,8-9H,13H2;1-9H3;1H;/q;;;+1/p-1

InChI key

ZVSLIOFJVMRWHJ-UHFFFAOYSA-M

General description

P(t-Bu)3 Pd G2 (Chloro[(tri-tert-butylphosphine)-2-(2-aminobiphenyl)] palladium(II)) is a second generation (G2) precatalyst containing a biphenyl-based ligand. Product participates in various palladium catalyzed cross-coupling reactions, C-C, C-N and C-O bond formation reactions, and Suzuki-Miyaura coupling reactions. It generates active Pd catalyst at room temperature in the presence of weak phosphate or carbonate bases. It has been proposed as an active catalyst for use in Stille reactions of aryl halides (bromides and chlorides).

Application

P(t-Bu)3 Pd G2 (Chloro[(tri-tert-butylphosphine)-2-(2-aminobiphenyl)] palladium(II), Pd/P(t-Bu)3) may be used as catalyst in the following studies:
  • Synthesis of sterically hindered biaryls (tetra-ortho-substituted), via cross-coupling reactions of aryl chlorides.
  • Stille cross-couplings reactions of aryl chloride.
  • Synthesis of chloropeptin I, via Stille cross-coupling reaction.
  • Heck reaction.
  • Negishi cross-coupling reactions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adam F Littke et al.
Journal of the American Chemical Society, 124(22), 6343-6348 (2002-05-30)
Pd/P(t-Bu)(3) serves as an unusually reactive catalyst for Stille reactions of aryl chlorides and bromides, providing solutions to a number of long-standing challenges. An unprecedented array of aryl chlorides can be cross-coupled with a range of organotin reagents, including SnBu(4).
Gregory C Fu
Accounts of chemical research, 41(11), 1555-1564 (2008-10-25)
Metal-catalyzed coupling reactions of aryl electrophiles with organometallics and with olefins serve as unusually effective tools for forming new carbon-carbon bonds. By 1998, researchers had developed catalysts that achieved reactions of aryl iodides, bromides, and triflates. Nevertheless, many noteworthy challenges

Articles

Buchwald precatalysts and ligands are bulky electron-rich dialkylbiaryl phospine-based catalysts and structurally related ligands that improve reactivity in Pd-catalyzed cross - coupling reactions

Buchwald precatalysts and ligands are bulky electron-rich dialkylbiaryl phospine-based catalysts and structurally related ligands that improve reactivity in Pd-catalyzed cross - coupling reactions

Buchwald precatalysts and ligands are bulky electron-rich dialkylbiaryl phospine-based catalysts and structurally related ligands that improve reactivity in Pd-catalyzed cross - coupling reactions

Buchwald precatalysts and ligands are bulky electron-rich dialkylbiaryl phospine-based catalysts and structurally related ligands that improve reactivity in Pd-catalyzed cross - coupling reactions

See All

Related Content

The Buchwald group has developed a series of highly active and versatile palladium precatalysts and biarylphosphine ligands used in cross-coupling reactions for the formation of C-C, C–N, C–O, C–F, C–CF3, and C–S bonds. The ligands are electron-rich, and highly tunable to provide catalyst systems with a diverse scope, high stability and reactivity. Furthermore, the new series of precatalysts are air-, moisture and thermally-stable and display good solubility in common organic solvents. The use of precatalysts ensures the efficient generation of the active catalytic species and allows one to accurately adjust the ligand:palladium ratio. The ligands, precatalysts and methodology developed in the Buchwald group are user friendly and have rendered previously difficult cross couplings reactions, much easier to achieve.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service