Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

922900

Sigma-Aldrich

GPhos Pd G6 TES

Synonym(s):

GPhos OAC precatalyst TES

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C47H70BrO4PPdSi
CAS Number:
Molecular Weight:
944.44
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.22

form

powder

Quality Level

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

SMILES string

[Pd]([PH](C5CCCCC5)(C4CCCCC4)c2c(ccc(c2c3c(cccc3C(C)C)C(C)C)C)C)(Br)c1ccc(cc1)C(=O)OCC[Si](C)(C)C

InChI

1S/C32H47P.C12H17O2Si.BrH.Pd/c1-22(2)28-18-13-19-29(23(3)4)31(28)30-24(5)20-21-25(6)32(30)33(26-14-9-7-10-15-26)27-16-11-8-12-17-27;1-15(2,3)10-9-14-12(13)11-7-5-4-6-8-11;;/h13,18-23,26-27H,7-12,14-17H2,1-6H3;5-8H,9-10H2,1-3H3;1H;

InChI key

TZDQPFRWRQKAKO-UHFFFAOYSA-N

Application

GPhos Pd G6 TES is a sixth generation (G6) Buchwald palladium precatalyst for cross-coupling reactions with demonstrated use for the room temperature coupling of primary amines with aryl halides.

Learn more about G6 Buchwald precatalysts

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elaine C Reichert et al.
Journal of the American Chemical Society, 145(6), 3323-3329 (2023-02-01)
We report a versatile and functional-group-tolerant method for the Pd-catalyzed C-N cross-coupling of five-membered heteroaryl halides with primary and secondary amines, an important but underexplored transformation. Coupling reactions of challenging, pharmaceutically relevant heteroarenes, such as 2-H-1,3-azoles, are reported in good-to-excellent
Development of an Aryl Amination Catalyst with Broad Scope Guided by Consideration of Catalyst Stability
Scott D, et al
Journal of the American Chemical Society, 142, 15027-15037 (2020)

Articles

Buchwald G6 precatalysts are oxidative addition complexes that are thermally, air, and moisture stable. The catalyst activation doesn’t require a base and generates innocuous byproducts.

Buchwald G6 precatalysts are oxidative addition complexes that are thermally, air, and moisture stable. The catalyst activation doesn’t require a base and generates innocuous byproducts.

Buchwald G6 precatalysts are oxidative addition complexes that are thermally, air, and moisture stable. The catalyst activation doesn’t require a base and generates innocuous byproducts.

Buchwald G6 precatalysts are oxidative addition complexes that are thermally, air, and moisture stable. The catalyst activation doesn’t require a base and generates innocuous byproducts.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service