Skip to Content
MilliporeSigma
All Photos(3)

Documents

294942

Sigma-Aldrich

N,N-Dicyclohexylmethylamine

97%

Synonym(s):

N-Methyldicyclohexylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H11)2NCH3
CAS Number:
Molecular Weight:
195.34
Beilstein:
2074991
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.49 (lit.)

bp

265 °C (lit.)

density

0.912 g/mL at 25 °C (lit.)

SMILES string

CN(C1CCCCC1)C2CCCCC2

InChI

1S/C13H25N/c1-14(12-8-4-2-5-9-12)13-10-6-3-7-11-13/h12-13H,2-11H2,1H3

InChI key

GSCCALZHGUWNJW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N,N-Dicyclohexylmethylamine has been employed as:
  • catalyst during O-phenylation of tertiary alcohol with organobismuth(V) compounds
  • base during Pd-catalyzed 5-endo-trig cyclization of 1-(o-bromophenyl)-2-methylprop-2-en-1-ol

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sriram Kumar et al.
Nature communications, 11(1), 2960-2960 (2020-06-13)
Nucleic acid-based materials enable sub-nanometer precision in self-assembly for fields including biophysics, diagnostics, therapeutics, photonics, and nanofabrication. However, structural DNA nanotechnology has been limited to substantially hydrated media. Transfer to organic solvents commonly used in polymer and peptide synthesis results
Copper (II)-catalyzed O-phenylation of alcohols with organobismuth (V) reagents.
Sakurai N, et al.
ARKIVOC (Gainesville, FL, United States), 7, 254-264 (2007)
Heck-type reactions of allylic alcohols: Part IV:(2-Substituted)-1-indanones via 5-endo-trig cyclizations.
Zawisza AM, et al.
J. Mol. Catal. A: Chem., 283(1), 140-145 (2008)

Articles

What is COgen? What is COware? What is the SyTracks COgenerator system? And more questions answered.

What is COgen? What is COware? What is the SyTracks COgenerator system? And more questions answered.

What is COgen? What is COware? What is the SyTracks COgenerator system? And more questions answered.

What is COgen? What is COware? What is the SyTracks COgenerator system? And more questions answered.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service