Skip to Content
MilliporeSigma
All Photos(1)

Documents

749087

Sigma-Aldrich

5-Thiazole boronic acid MIDA ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9BN2O4S
Molecular Weight:
240.04
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

solid

mp

172-178 °C

storage temp.

2-8°C

SMILES string

CN1CC(=O)OB(OC(=O)C1)c2cncs2

InChI

1S/C8H9BN2O4S/c1-11-3-7(12)14-9(15-8(13)4-11)6-2-10-5-16-6/h2,5H,3-4H2,1H3

InChI key

JHLBEKAIQFBSNV-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service