Skip to Content
MilliporeSigma
All Photos(1)

Documents

281077

Sigma-Aldrich

Trimethyloxonium tetrafluoroborate

95%

Synonym(s):

Trimethyloxonium fluoroborate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3O(BF4)
CAS Number:
Molecular Weight:
147.91
Beilstein:
3597303
EC Number:
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

solid

storage temp.

−20°C

SMILES string

C[O+](C)C.F[B-](F)(F)F

InChI

1S/C3H9O.BF4/c1-4(2)3;2-1(3,4)5/h1-3H3;/q+1;-1

InChI key

CZVZBKHWOFJNCR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Trimethyloxonium tetrafluoroborate can be used as a methylating agent for the methylation of hydroxyl/carboxyl functional groups. It is capable of methylating polyfunctional carboxylic acids. It is also used as a catalyst for the polymerization of cyclic sulfides and ethers.

Application

Reagent for the methylation of hydroxyl groups recently used in a complex, multistep synthesis directed towards spirastrellolide, a marine natural product.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toward the total synthesis of spirastrellolide A. Part 2: Conquest of the northern hemisphere.
Alois Fürstner et al.
Angewandte Chemie (International ed. in English), 45(33), 5510-5515 (2006-08-15)
H M Liebich et al.
Journal of chromatography. A, 843(1-2), 237-245 (1999-07-10)
Trimethyloxonium tetrafluoroborate (TMO) is applied as derivatising reagent to transform urinary organic acids into their methyl esters. The method is suggested as an alternative to the use of diazomethane which is carcinogenic and explosive. In contrast to other methods avoiding
S Chericoni et al.
Journal of analytical toxicology, 35(4), 193-198 (2011-04-26)
The present work describes the validation of a novel aqueous in situ derivatization procedure with trimethyloxonium tetrafluoroborate (TMO) as methylating agent for the simultaneous, quantitative analysis of Δ(9)-tetrahydrocannabinol (THC) and 11-nor-Δ(9)-tetrahydrocannabinol carboxylic acid (THC-COOH) in human urine. The derivatizing agent
Marco Pacenti et al.
Biomedical chromatography : BMC, 22(10), 1155-1163 (2008-05-29)
A method for the determination of the organic acids directly in the urine employing derivatization with trimethyloxonium tetrafluoroborate as a methylating agent and sequential extraction by head space and direct immersion/solid phase microextraction is reported. Furoic acid, hippuric acid, methylhippuric
H M Liebich et al.
Journal of chromatography. B, Biomedical sciences and applications, 713(2), 427-432 (1998-09-24)
We developed a new sample preparation method for profiling organic acids in urine by GC or GC-MS. The method includes derivatisation of the organic acids directly in the aqueous urine using trimethyloxonium tetrafluoroborate as a methylating agent, extraction of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service