Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

571350

Sigma-Aldrich

1-Phenylvinylboronic acid

95%

Synonym(s):

α-Methylene-α-tolueneboronic acid, α-Phenyl vinylboronic acid, (1-Phenylethenyl)boronic acid, Styrene α-boronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C=CH2B(OH)2
CAS Number:
Molecular Weight:
147.97
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

mp

125 °C (dec.) (lit.)

functional group

phenyl

storage temp.

2-8°C

SMILES string

OB(O)C(=C)c1ccccc1

InChI

1S/C8H9BO2/c1-7(9(10)11)8-5-3-2-4-6-8/h2-6,10-11H,1H2

InChI key

YJCPVMYUISTDKG-UHFFFAOYSA-N

Application

Reactant involved in:
  • Halogenation of alkynes for preparation of enol esters
  • Liebeskind-Srogl cross-coupling
  • Homocoupling of arylboronic acids for synthesis of biaryls
  • Iterative cross-coupling of boronate building blocks
  • Enantioselective hydrogenation of hydroxyphenyl styrenes
  • Samarium diiodide-mediated cyclization for synthesis of substituted benzannulated cyclooctanol derivatives
  • Suzuki-Miyaura cross-coupling for synthesis of C-6-substituted uridine phosphonates

Other Notes

Contains varying amounts of anhydride

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service