Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

731234

Sigma-Aldrich

α-Tocopherol (phenyl-5,7-dimethyl-d6)

≥98 atom % D, ≥98% (CP)

Synonym(s):

Vitamin E (ring-5,7-dimethyl-d6)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29D6H44O2
CAS Number:
Molecular Weight:
436.74
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

isotopic purity

≥98 atom % D

Assay

≥98% (CP)

form

liquid

technique(s)

mass spectrometry (MS): suitable

color

yellow to brown

bp

200-220 °C/1 mmHg (lit.)

density

0.963 g/mL at 25 °C

mass shift

M+6

storage temp.

−20°C

SMILES string

OC1=C(C([2H])([2H])[2H])C(C)=C(O[C@@](CCC[C@H](C)CCC[C@H](C)CCCC(C)C)(C)CC2)C2=C1C([2H])([2H])[2H]

InChI

1S/C29H50O2/c1-20(2)12-9-13-21(3)14-10-15-22(4)16-11-18-29(8)19-17-26-25(7)27(30)23(5)24(6)28(26)31-29/h20-22,30H,9-19H2,1-8H3/t21-,22-,29-/m1/s1/i5D3,7D3

InChI key

GVJHHUAWPYXKBD-BCHQTGRQSA-N

Related Categories

Application

α-Tocopherol (phenyl-5,7-dimethyl-d6) is also known as vitamin E (ring-5,7-dimethyl-d6) that can be used as an internal standard. It is utilized to determine α-tocopherol (α-T) concentration in different biopharmaceutical samples using liquid chromatography-diode array detector on-line ESI-MS/MS.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

230.0 °F

Flash Point(C)

110 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ultrafast determination of vitamin E using LC-ESI-MS/MS for preclinical development of new nutraceutical formulations
Villaseca-Gonzalez N, et al.
Bioanalysis, 10(4), 215-227 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service