Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

790427

Sigma-Aldrich

Iron-57Fe

95 atom %, 99.9% (trace metals analysis)

Synonym(s):

Iron-57

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
57Fe
CAS Number:
Molecular Weight:
56.94
MDL number:
UNSPSC Code:
12141721
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

95 atom %

Quality Level

Assay

99.9% (trace metals analysis)

form

solid

mass shift

M+1

storage temp.

room temp

SMILES string

[57Fe]

InChI

1S/Fe/i1+1

InChI key

XEEYBQQBJWHFJM-OUBTZVSYSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eduard V Bocharov et al.
Biochemistry, 56(12), 1697-1705 (2017-03-16)
The epidermal growth factor receptor (EGFR) family is an important class of receptor tyrosine kinases, mediating a variety of cellular responses in normal biological processes and in pathological states of multicellular organisms. Different modes of dimerization of the human EGFR

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service